Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Coumaranone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Benzo[b]furan-2(3H)-oneBenzofuran-2(3H)-one

A18617-03 A18617-14 A18617-06 A18617-06 A18617-14 A18617-03
AAA18617-06EA 137.99 USD InStock
AAA18617-06 AAA18617-14 AAA18617-03 CAAAA18617-06 CAAAA18617-03 CAAAA18617-14
2-Coumaranone 99%
2-Coumaranone


Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 248…250 °C
Melting Pt: 47…52 °C
Density: 1,22
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005856
CAS Number: 553-86-6
EINECS: 209-052-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty