Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bisabolene mixture of isomers 96% (sum of isomers)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(1,5-Dimethyl-4-hexenylidene)-1-methylcyclohexene

A18724-09 A18724-18 A18724-18 A18724-09
AAA18724-18EA 173.99 USD InStock
CAAAA18724-09 AAA18724-18 CAAAA18724-18 AAA18724-09
Bisabolene mixture of isomers 96% (sum of isomers)
Bisabolene


Formula: C₁₅H₂₄
MW: 204.36 g/mol
Density: 0.89
Storage Temperature: Ambient
MDL Number: MFCD00129080
CAS Number: 495-62-5
EINECS: 207-805-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty