Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Fluoreneacetic acid ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(9H-Fluoren-9-yl)acetic acid

A18772-03 A18772-06 A18772-06 A18772-03 A18772-14
AAA18772-03EA 77.89 USD InStock
AAA18772-03 CAAAA18772-03 CAAAA18772-14 CAAAA18772-06 AAA18772-06
9-Fluoreneacetic acid ≥99.5%
9-Fluoreneacetic acid


Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00013262
CAS Number: 6284-80-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty