Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Methylvaleric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-3-Methylpentanoic acid

A18886-22 A18886-06 A18886-06 A18886-14 A18886-14 A18886-22
AAAA18886-22EA 349.99 USD InStock
CAAAA18886-14 CAAAA18886-06 AAAA18886-22 AAA18886-14 CAAAAA18886-22 AAA18886-06
(±)-3-Methylvaleric acid 99%
(±)-3-Methylvaleric acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 196…198 °C
Melting Pt: –42 °C
Density: 0.930
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002729
CAS Number: 105-43-1
EINECS: 203-297-7
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty