Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Valine 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-Valine H-D-Val-OH

A18894-14 A18894-06 A18894-22 A18894-14 A18894-22 A18894-06
AAA18894-06EA 37.49 USD InStock
AAA18894-06 CAAAA18894-06 CAAAA18894-14 AAA18894-22 CAAAA18894-22 AAA18894-14
D-(-)-Valine 98+%
D(-)-Valine


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty