Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylcaproic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Methylhexanoic acid

A19012-18 A19012-18 A19012-09 A19012-09
AAA19012-09EA 81.09 USD InStock
CAAAA19012-18 AAA19012-09 AAA19012-18 CAAAA19012-09
5-Methylcaproic acid 98%
5-Methylcaproic acid


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 216 °C
Density: 0.91
Flash Pt: 102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036494
CAS Number: 628-46-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty