Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-β-Citronellol 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: β-Citronellol3,7-Dimethyloct-6-en-1-olCitronellol(±)-CitronellolDL-Citronellol

A19016-36 A19016-22 A19016-22 A19016-36
AAA19016-22EA 44.79 USD InStock
CAAAA19016-36 AAA19016-22 AAA19016-36 CAAAA19016-22
(±)-β-Citronellol 95%
(±)-β-Citronellol


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 221…222 °C
Melting Pt: < –20 °C
Density: 0.858
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
ADR: 9,III
Merck Index: 13,02354
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty