Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-2-pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning

A19090-22 A19090-14 A19090-22 A19090-14
AAA19090-14EA 147.99 USD InStock
AAA19090-14 CAAAA19090-14 AAA19090-22 CAAAA19090-22
2-Methyl-2-pentanol 99%
2-Methyl-2-pentanol


Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 120…122 °C
Melting Pt: –103 °C
Density: 0.810
Flash Pt: 30 °C(86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004485
CAS Number: 590-36-3
EINECS: 209-681-0
UN: 2560
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty