Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-ethylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 6-Ethyl-2-pyridylamine 6-Ethylpyridin-2-amine

A19146-09 A19146-09
AAA19146-09EA 241.99 USD InStock
CAAAA19146-09 AAA19146-09
2-Amino-6-ethylpyridine 97%
2-Amino-6-ethylpyridine


Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00218265
CAS Number: 21717-29-3
EINECS: 244-540-7
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty