Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-4-fluorobenzylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A19280-06 A19280-06 A19280-14 A19280-14
AAA19280-06EA 82.49 USD InStock
AAA19280-06 CAAAA19280-06 CAAAA19280-14 AAA19280-14
3-Bromo-4-fluorobenzylamine hydrochloride 98%
3-Bromo-4-fluorobenzylamine hydrochloride


Formula: C₇H₈BrClFN
MW: 240.50 g/mol
Melting Pt: 214…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00143094
CAS Number: 202865-68-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty