Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-1-iodo-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Chloro-4-iodonitrobenzene

A19340-03 A19340-14 A19340-06 A19340-06 A19340-14 A19340-03
AAAA19340-14EA 559.99 USD InStock
AAAA19340-14 AAA19340-06 AAA19340-03 CAAAA19340-03 CAAAA19340-06 CAAAAA19340-14
2-Chloro-1-iodo-4-nitrobenzene 98%
2-Chloro-1-iodo-4-nitrobenzene


Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00024589
CAS Number: 41252-96-4
EINECS: 255-278-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty