Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-o-anisaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromo-2-methoxybenzaldehyde5-Bromo-2-anisaldehyde

A19396-14 A19396-36 A19396-22 A19396-36 A19396-22 A19396-14
AAA19396-22EA 149.99 USD InStock
AAA19396-22 CAAAAA19396-36 CAAAA19396-22 CAAAA19396-14 AAAA19396-36 AAA19396-14
5-Bromo-o-anisaldehyde 98+%
5-Bromo-o-anisaldehyde


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty