Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Sulfobenzoic acid monopotassium salt 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Sulphobenzoic acid potassium salt4-Sulfobenzoic acid potassium salt

A19432-30 A19432-18 A19432-18 A19432-30
AAA19432-30EA 190.99 USD InStock
AAA19432-30 CAAAA19432-18 CAAAA19432-30 AAA19432-18
4-Sulfobenzoic acid monopotassium salt 95%
4-Sulphobenzoic acid monopotassium salt


Formula: C₇H₅KO₅S
MW: 240.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007509
CAS Number: 5399-63-3
EINECS: 226-435-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty