Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-methyl-4-pentenoate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-4-pentenoic Acid Ethyl Ester

A19526-06 A19526-06 A19526-22 A19526-22 A19526-14 A19526-14
AAA19526-22EA 211.99 USD InStock
CAAAA19526-06 AAA19526-22 CAAAA19526-14 CAAAA19526-22 AAA19526-06 AAA19526-14
Ethyl-2-methyl-4-pentenoate 98%
Ethyl-2-methyl-4-pentenoate


Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Boiling Pt: 153…155 °C
Density: 0.880
Flash Pt: 40 °C(104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010235
CAS Number: 53399-81-8
EINECS: 258-520-0
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty