Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylglycine 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (Dimethylamino)acetic acid2-(Dimethylamino)acetic acid

A19670-14 A19670-06 A19670-14 A19670-06 A19670-22 A19670-22
AAA19670-22EA 889.99 USD InStock
AAA19670-22 CAAAA19670-14 CAAAA19670-06 CAAAA19670-22 AAA19670-06 AAA19670-14
N,N-Dimethylglycine 98+%
N,N-Dimethylglycine


Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty