Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexanecarbonyl chloride 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclohexanecarboxylic acid chloride Cyclohexanecarboxylic chloride

A19824-22 A19824-14 A19824-14 A19824-22
AAA19824-22EA 77.49 USD InStock
AAA19824-22 AAA19824-14 CAAAA19824-14 CAAAA19824-22
Cyclohexanecarbonyl chloride 97+%
Cyclohexanecarbonyl chloride


Formula: C₇H₁₁ClO
MW: 146.62 g/mol
Boiling Pt: 183…185 °C
Melting Pt: 29 °C
Density: 1.088
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001456
CAS Number: 2719-27-9
EINECS: 220-322-7
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty