Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-mercaptobenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Chloro-2-benzimidazolethiol

A19852-14 A19852-06 A19852-14 A19852-06
AAA19852-14EA 334.99 USD InStock
AAA19852-14 CAAAA19852-06 AAA19852-06 CAAAA19852-14
5-Chloro-2-mercaptobenzimidazole 98%
5-Chloro-2-mercaptobenzimidazole


Formula: C₇H₅ClN₂S
MW: 184.64 g/mol
Melting Pt: 298…302 °C
Storage Temperature: Ambient
MDL Number: MFCD01658762
CAS Number: 25369-78-2
EINECS: 246-903-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty