Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorophenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,5-Dichlorophenylisocyantae

A19991-14 A19991-06 A19991-06 A19991-14
AAA19991-06EA 78.29 USD InStock
CAAAA19991-14 CAAAA19991-06 AAA19991-06 AAA19991-14
3,5-Dichlorophenyl isocyanate 97%
3,5-Dichlorophenyl isocyanate


Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 242…244 °C
Melting Pt: 28…32 °C
Density: 1.380
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013859
CAS Number: 34893-92-0
EINECS: 252-276-9
UN: 2250
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty