Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Malonic acid ≥99.5%, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3-Propanedioic acid

031715-22 031715-A4 031715-36
AAA31715-22EA 68.94 USD InStock
AAA31715-22 AAA31715-A4 AAA31715-36
Malonic acid ≥99.5%, Reagent Grade
Malonic acid


Formula: C₃H₄O₄
MW: 104.06 g/mol
Boiling Pt: 387 °C (1013 hPa)
Melting Pt: 132…136 °C (dec.)
Density: 1,62
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0
Merck Index: 13,05732
Order Now
Specification Test Results
Form Crystalline

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty