Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oleic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (9Z)-9-Octadecenoic acid

031997-06 31997-22 31997.22 031997-14 31997-14 31997-06
AAA31997-22EA 988.99 USD InStock
AAA31997-22 CAAA31997-14 CAAAA31997-22 CAAA31997-06 AAA31997-14 AA31997-06
Oleic acid 99%
Oleic acid


Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286 °C (100 mmHg)
Melting Pt: 13…14 °C
Density: 0.887
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/57/2298957.jpg
Order Now
Specification Test Results
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty