Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Millerite Nickel sulphide

35661-09 35661-30 35661.09 035661-18 35661-30 35661-18
AA35661-18EA 417.99 USD InStock
CAAAA35661-09 CAAA35661-18 AA35661-18 CAAA35661-30 AA35661-30 AAA35661-09
Nickel sulfide ≥99.9% (metals basis)
Nickel sulfide


Formula: NiS
MW: 240.21 g/mol
Melting Pt: 797 °C
Storage Temperature: Ambient
MDL Number: MFCD00016266
CAS Number: 16812-54-7
EINECS: 234-829-6
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Form -200 Mesh Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty