Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN

036423-M1 036423-AP 036423-K2 036423-K7 036423-M6
AA36423-K7EA 324.41 USD InStock
AA36423-K7 AA36423-M6 AA36423-M1 AAA36423-K2 AA36423-AP
Acetonitrile ≥99.5% ACS
Acetonitrile


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 80…82 °C
Melting Pt: –48…–45 °C
Density: 0.786
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Form Liquid
Evaporation residue 0.005%
Titratable acid 8µeq/g
Titratable base 0.06µg/g
H2O 0.3%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty