Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3'-Diethylthiadicarbocyanine iodide ≥99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DiSC2(5)3-Ethyl-2-[5-(3-ethyl-2(3H)-benzothiazolylidene)-1,3-pentadienyl]benzothiazolium iodideDTDCI

43851-04 43851.02 43851.09 43851.04 43851-09 43851-02
AAA43851-09EA 396.99 USD InStock
AAA43851-09 CAAAA43851-02 CAAAA43851-04 AAA43851-02 CAAAA43851-09 AAA43851-04
3,3'-Diethylthiadicarbocyanine iodide ≥99%
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)]
Applications: As a solvent soluble laser dye


Formula: C₂₃H₂₃N₂S₂I
MW: 518.48 g/mol
Melting Pt: 248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00074829
CAS Number: 514-73-8
EINECS: 208-186-7
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty