Synonyms:
N,N'-(Disulfanediylbis(ethane-2,1-diyl))diacrylamide
, N,N'-(Disulphanediylbis(ethane-2,1-diyl))diacrylamide
44132.14
44132.03
44132.06
44132-14
44132-03
44132-06
AAA44132-06EA
322.99
USD
InStock
CAAAA44132-14
CAAAA44132-03
CAAAA44132-06
AAA44132-06
AAA44132-14
AAA44132-03
N,N'-Bis(acryloyl)cystamine ≥97%
N,N'-Bis(acryloyl)cystamine
Powder, Packaged under argon
Applications: Used as a reversible cross-linker for polyacrylamide gels