Synonyms:
Fullerene carbon nanotube
44192.06
044192-03
44192-03
44192-06
AAA44192-03EA
215.99
USD
InStock
AAA44192-03
AAA44192-06
CAAAA44192-06
CAAAA44192-03
Fullerene nanotube multi-walled, ground core 20-50 nm OD, Length 5-20 micron
Fullerene nanotube
Powder