Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)zirconium dihydride 98%

Supplier: Thermo Scientific Chemicals
Danger

44377.03 44377.06 44377-06 44377-03
AAA44377-06EA 222.99 USD InStock
AAA44377-06 CAAAA44377-06 CAAAA44377-03 AAA44377-03
Bis(cyclopentadienyl)zirconium dihydride 98%
Bis(cyclopentadienyl)zirconium dihydride
Powder

Hydrogenation catalyst for alkenes and alkynes


Formula: C₁₀H₁₂Zr
MW: 223.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04126436
CAS Number: 37342-98-6
UN: 3131
ADR: 4.3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty