Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Poly(styrene-divinylbenzene), aminomethylated, cross-linked with 1% DVB 100-200 mesh

Supplier: Thermo Scientific Chemicals

44392-22 44392.06 44392-22 44392-06 44392-14 44392.14
AAA44392-06EA 62.59 USD InStock
AAA44392-06 CAAAA44392-06 AAA44392-22 CAAAA44392-14 CAAAA44392-22 AAA44392-14
Poly(styrene-divinylbenzene), aminomethylated, cross-linked with 1% DVB 100-200 mesh
Poly(styrene-divinylbenzene), aminomethylated
Powder

Applications: Formation of amide groups using Boc strategy; as a scavenger resin


Storage Temperature: Ambient
MDL Number: MFCD00146442
CAS Number: 89551-24-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty