Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)

Supplier: Thermo Scientific Chemicals
Warning

44440.22 44440-14 44440-22 44440.14
AAA44440-14EA 248 USD InStock
AAA44440-14 AAA44440-22 CAAAA44440-22 CAAAA44440-14
Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)
Barium oxide


Formula: BaO
MW: 153.33 g/mol
Boiling Pt: ∼2000 °C (1013 hPa)
Melting Pt: 1923 °C
Density: 5.7
Storage Temperature: Ambient
MDL Number: MFCD00003453
CAS Number: 1304-28-5
EINECS: 215-127-9
UN: 1884
ADR: 6.1,III
Merck Index: 12,01015
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty