Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver perrhenate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Silver(I) perrhenate

44480-06 44480.03 44480-03 44480.06
AAA44480-03EA 90.49 USD InStock
CAAAA44480-03 AAA44480-03 AAA44480-06 CAAAA44480-06
Silver perrhenate ≥99.99% (metals basis)
Silver perrhenate


Formula: AgO₄Re
MW: 358.07 g/mol
Melting Pt: 430 °C
Density: 7.05
Storage Temperature: Ambient
MDL Number: MFCD00014144
CAS Number: 20654-56-2
EINECS: 232-042-2
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty