Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicic acid potassium salt, anhydrous

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Potassium silicate

44493-22 44493-A3 44493.22 044493.A3 44493-36 44493.36
AAA44493-36EA 90.39 USD InStock
CAAAA44493-36 CAAAA44493-A3 CAAAA44493-22 AAA44493-36 AAA44493-A3 AAA44493-22
Silicic acid potassium salt, anhydrous
Silicic acid potassium salt
Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.

Caution: Moisture sensitive. Incompatible with strong oxidizing agents.


Storage Temperature: Ambient
MDL Number: MFCD00145152
CAS Number: 1312-76-1
EINECS: 215-199-1
Order Now
Specification Test Results
Assay (unspecified) 28.0 - 28.8% K₂O

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty