Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl)silane

44543.04 44543-04 44543.09 44543-09
AAA44543-04EA 72.99 USD InStock
AAA44543-04 CAAAA44543-04 CAAAA44543-09 AAA44543-09
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane 97%
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Liquid


Formula: C₁₀H₁₀ClF₁₃Si
MW: 440.71
Boiling Pt: 189…191 °C
Density: 1,461
Flash Pt: 52 °C (180 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042359
CAS Number: 102488-47-1
UN: 2986
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty