Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Soda lime ACS, with indicator

Supplier: Thermo Scientific Chemicals
Danger

044697.A4 44697-A4 44697-36 44697.36
AAA44697-A4EA 333.11 USD InStock
AAA44697-A4 AAA44697-36 CAAAA44697-A4 CAAAA44697-36
Soda lime ACS, with indicator
Soda lime


Storage Temperature: Ambient
MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty