Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2,Specpure®, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
44789-AE 044789-AE
AAA44789-AEEA 257.16 USD InStock
CAAAA44789-AE AAA44789-AE
ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2,Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards
These standards are applicable for the determination of elements as dissolved elements in ground waters, surface waters, and drinking water, and total recoverable elements in these waters as well as wastewaters, sludges, and soils samples.

Matrix: 5% HNO3
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD03792350

Concentration: Bi 10 µg/mL ±2%; In 10 µg/mL ±2%; Lu 10 µg/mL ±2%; Sc 10 µg/mL ±2%; Tb 10 µg/mL ±2%; Y 10 µg/mL ±2%; ₆Li 10 µg/mL ±2%

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty