Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium malonate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Malonic acid disodium saltSodium malonate dibasic

44841.14 44841-22 44841-14 44841.22
AAA44841-22EA 98.29 USD InStock
AAA44841-22 AAA44841-14 CAAAA44841-22 CAAAA44841-14
Disodium malonate 99%
Disodium malonate


Formula: C₃H₂Na₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002708
CAS Number: 141-95-7
EINECS: 205-514-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty