Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentaethylene glycol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

A10133-30 A10133-18 A10133-09 A10133-30 A10133-18 A10133-09
AAAA10133-30EA 1851.99 USD InStock
AAAA10133-30 AAAA10133-18 CAAAAA10133-09 CAAAAA10133-18 CAAAAA10133-30 AAAA10133-09
Pentaethylene glycol 98+%
Pentaethylene glycol


Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Boiling Pt: 338…340 °C
Density: 1,126
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty