Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A10207-14 A10207-22 A10207-06 A10207-06 A10207-14 A10207-22
AAAA10207-22EA 407.99 USD InStock
AAAA10207-22 CAAAAA10207-22 AAA10207-06 AAA10207-14 CAAAA10207-14 CAAAA10207-06
4-Amino-3-hydroxybenzoic acid 98%
4-Amino-3-hydroxybenzoic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty