Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) sulfide Pb ≥ 82% (≥82% Pb)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Lead sulphideLead sulfide

A10268-0E A10268-0E A10268-36 A10268-36
AAAA10268-0EEA 270.99 USD InStock
AAAA10268-0E CAAAAA10268-36 CAAAAA10268-0E AAAA10268-36
Lead(II) sulfide Pb ≥ 82% (≥82% Pb)
Lead (II) sulphide


Formula: ̍PbS
MW: 239.27 g/mol
Melting Pt: 1114 °C
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: "9",III
Merck Index: 12,05445
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty