Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Bromocyclohexene 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Bromocyclohexene

Stabilization: Stabilised with propylene oxide 300-1000 ppm

A10370-22 A10370-14 A10370-06 A10370-14 A10370-06 A10370-22
AAAA10370-14EA 181.99 USD InStock
CAAAAA10370-22 CAAAAA10370-06 AAAA10370-14 AAAA10370-22 CAAAAA10370-14 AAAA10370-06
(±)-3-Bromocyclohexene 95% stabilized
(±)-3-Bromocyclohexene


Formula: C₆H₉Br
MW: 161.04 g/mol
Boiling Pt: 64…65 °C/15 mm
Density: 1.400
Flash Pt: 54 °C(129 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013775
CAS Number: 1521-51-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty