Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA (ethylenediamine tetraacetic acid) 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: EDTAEdetic AcidEthylenediaminetetraacetic acid

A10713-0I A10713-0B A10713-36 A10713-36 A10713-0I A10713-0B
AAA10713-0BEA 85.4 USD InStock
AAA10713-0B CAAAAA10713-36 CAAAA10713-0I AAA10713-0I AAAA10713-36 CAAAA10713-0B
EDTA (ethylenediamine tetraacetic acid) 99%
EDTA (Ethylenediamine tetraacetic acid)


Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/41/2297241.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty