Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butylbenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: n-Butylbenzene

A10910-30 A10910-0B A10910-18 A10910-30 A10910-0B A10910-18
AAAA10910-0BEA 567.99 USD InStock
CAAAAA10910-0B AAAA10910-0B AAAA10910-30 CAAAAA10910-30 CAAAAA10910-18 AAAA10910-18
Butylbenzene 99%
Butylbenzene


Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 181…183 °C
Melting Pt: –88 °C
Density: 0.860
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009463
CAS Number: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: "3",III
Merck Index: 13,01548
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty