Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexafluorotitanate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium hexafluorotitanate(IV)

A10915-36 A10915-0E A10915-22 A10915-0E A10915-22 A10915-36
AAAA10915-0EEA 197.99 USD InStock
AAAA10915-0E CAAAAA10915-0E AAAA10915-36 CAAAAA10915-36 AAAA10915-22 CAAAAA10915-22
Potassium hexafluorotitanate 97%
Potassium hexafluorotitanate


Formula: F₆K₂Ti
MW: 240.07 g/mol
Boiling Pt: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00011380
CAS Number: 16919-27-0
EINECS: 240-969-9
UN: 3288
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty