Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitroquinaldine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-6-nitroquinoline

A10945-03 A10945-06 A10945-14 A10945-03 A10945-14 A10945-06
AAA10945-03EA 81.39 USD InStock
CAAAA10945-06 CAAAAA10945-14 CAAAA10945-03 AAA10945-03 AAAA10945-14 AAA10945-06
6-Nitroquinaldine 98%
6-Nitroquinaldine


Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00051736
CAS Number: 613-30-9
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty