Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Furfuryl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Furfurol

A10968-18 A10968-18 A10968-0B A10968-0B A10968-30 A10968-30
AAAA10968-30EA 37.63 USD InStock
AAAA10968-30 CAAAAA10968-30 AAAA10968-18 CAAAAA10968-18 AAAA10968-0B CAAAAA10968-0B
Furfuryl alcohol 98%
Furfuryl alcohol


Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 170…171 °C
Melting Pt: –31 °C
Density: 1.129
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: "6.1",III
Merck Index: 13,04326
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty