Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroiodomethane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Stabilization: Stabilised with copper

A11032-09 A11032-18 A11032-30 A11032-09 A11032-30 A11032-18
AAAA11032-30EA 581.99 USD InStock
AAAA11032-30 CAAAAA11032-30 CAAAAA11032-09 AAAA11032-09 AAAA11032-18 CAAAAA11032-18
Chloroiodomethane 98% stabilized
Chloroiodomethane


Formula: CH₂ClI
MW: 176.38 g/mol
Boiling Pt: 108…109 °C
Density: 2.416
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty