Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Aminoisobutane

A11135-0D A11135-0F A11135-0D A11135-AP A11135-0F A11135-AP
AAAA11135-0FEA 120.99 USD InStock
AAAA11135-0F CAAAAA11135-AP AAAA11135-AP CAAAAA11135-0F AAAA11135-0D CAAAAA11135-0D
tert-Butylamine 98%
tert-Butylamine


Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 44…46 °C
Melting Pt: –67 °C
Density: 0.696
Flash Pt: -38 °C(-36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: "8",II
Merck Index: 13,01544
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty