Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caprylic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Octoic acid Octanoic acid

A11149-0F A11149-AC A11149-0F A11149-0D A11149-AC A11149-AP A11149-AP A11149-0D
AAA11149-APEA 56.38 USD InStock
AAA11149-AP CAAAA11149-0D CAAAAA11149-AC CAAAA11149-AP AAA11149-0F AAAA11149-AC AAA11149-0D CAAAA11149-0F
Caprylic acid 98+%
Caprylic acid


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 236…238 °C
Melting Pt: 16…17 °C
Density: 0.910
Flash Pt: 132 °C(269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: "8",III
Merck Index: 13,01771
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty