Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyadenosine monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

A11166-03 A11166-14 A11166-06 A11166-06 A11166-14 A11166-03
AAA11166-03EA 39.39 USD InStock
CAAAAA11166-14 AAA11166-03 AAA11166-06 CAAAA11166-06 AAAA11166-14 CAAAA11166-03
2'-Deoxyadenosine monohydrate 99%
2'-Deoxyadenosine monohydrate


Formula: C₁₀H₁₃N₅O₃·1H₂O
MW: 269.26 g/mol (251.24 anhy)
Melting Pt: 186…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149364
CAS Number: 16373-93-6
EINECS: 213-488-7
Order Now
Specification Test Results
Optical rotation -25° (c=0.5 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty