Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-m-toluonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methyl-4-nitrobenzonitrile

A11301-06 A11301-03 A11301-14 A11301-06 A11301-09 A11301-03 A11301-09 A11301-14
AAA11301-06EA 108.99 USD InStock
CAAAAA11301-09 AAA11301-06 CAAAA11301-03 CAAAA11301-14 CAAAA11301-06 AAA11301-03 AAA11301-14 AAAA11301-09
4-Nitro-m-toluonitrile 97%
4-Nitro-m-toluonitrile


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00017615
CAS Number: 96784-54-2
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty