Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate, dried 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: Dipotassium hydrogen phosphatePotassium hydrogen phosphate

A11321-0B A11321-30 A11321-0I A11321-30 A11321-0I A11321-0B
AAA11321-30EA 65.81 USD InStock
CAAAA11321-0B AAA11321-30 CAAAA11321-30 AAA11321-0B CAAAAA11321-0I AAAA11321-0I
di-Potassium hydrogen phosphate, dried 98+%
di-Potassium hydrogen phosphate


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty