Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A11396-18 A11396-30 A11396-09 A11396-18 A11396-09 A11396-30
AAA11396-18EA 145.99 USD InStock
AAA11396-18 CAAAA11396-09 AAA11396-09 CAAAAA11396-30 AAAA11396-30 CAAAA11396-18
3-Fluorobenzaldehyde 97%
3-Fluorobenzaldehyde


Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 173 °C
Density: 1.174
Flash Pt: 56 °C(132 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty